Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Human FKBP5 (aa 394-407) Synthetic Peptide
Synthetic Peptide
Supplier: Invitrogen™ PEP078
Description
Human FKBP51 (residues 394-407) Synthetic Peptide, PEP-078, from Invitrogen.
PEP-078 is a 14 amino acid synthetic peptide whose sequence corresponds to residues 394-407 of human FKBP51 with a carboxy terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): C (394) -Q-K-K-A-K-E-H-N-E-R-D-R-R (407)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human FKBP51, catalog # PA1-020. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
2289 | |
purified | |
Human FKBP5 (aa 394-407) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
Q13451 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
FKBP5 | |
No preservative | |
50 μg | |
C(394) -Q - K - K - A - K - E - H - N - E - R - D - R - R(407) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title