Learn More
Clontech Labs 3P HUMAN ADIPOSE TISSUE RNA
Supplier:  Clontech Labs 3P 636558
Encompass
Human Adipose Tissue Total RNA, Size: 10microgram, It is isolated by a modified guanidinium thiocyanate method
Catalog No. NC9160098