Learn More
STEMCELL Technologies Human Peripheral Blood Pan-T Cells, Frozen, Size: 4 x 10^7 cells
Supplier: STEMCELL Technologies 70024.1
Choose ready-to-use, ethically sourced, primary human pan T cells. With personalized service, custom products, flexible delivery times, and the option to reserve entire lots to prescreen cells for applications, we help you get the cells you need. Isolated from peripheral blood mononuclear cells using negative immunomagnetic separation and cryopreserved in CryoStorCS10 medium, pan T cells consist of CD4 and CD8 T cells and some gamma/delta T cell subsets. Cells are obtained using Institutional Review Board (IRB)-approved consent forms and protocols. Additional documentation and high-resolution HLA typing (Class I and Class II alleles and CMV status) are available upon request. Acid-citrate-dextrose solution A (ACDA) is added during collection as an anticoagulant. Contact techsupport@stemcell.com for donor specifications (BMI category, smoking status, etc.). Donors are screened for HIV-1, HIV-2, hepatitis B, and hepatitis C. Certain products are only available in select territories.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.