Learn More
STEMCELL Technologies Human Peripheral Blood Leukopak, Fresh, Size: 9 x 10^9 cells minimum
Supplier: STEMCELL Technologies 70500
Obtain large numbers of single-donor mononuclear cells (PBMCs) from normal peripheral blood and reduce the time and reagents required for your downstream cell isolation. Enjoy personalized service, custom products, flexible delivery times, and multiple size options. Leukopaks are highly concentrated, low-volume apheresis collections from normal donors, with Institutional Review Board (IRB)-approved consent forms and protocols. Each full-sized leukopak is produced from ~2 - 3x blood volumes using the Spectra Optia Apheresis System (unless otherwise requested), with ACDA as the anticoagulant. High-resolution HLA typing (Class I and Class II alleles and CMV status) are available upon request. Contact techsupport@stemcell.com for more information, including donor selection or information (BMI category, smoking status, ethnicity, etc.). Donors are recallable and are screened for HIV-1, HIV-2, hepatitis B, and hepatitis C. Certain products are only available in select territories.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.