Learn More
Vector Laboratories Avidin D Horseradish Peroxidase (Av-HRP) Concentrate For IHC 5 mg
Supplier: Vector Laboratories A20045
Horseradish Peroxidase Avidin D (HRP Avidin D) is produced by a coupling procedure that preserves the high specific activity of the peroxidase. Using biotinylated primary or secondary intermediates and peroxidase-labeled Avidin D, antigens can be localized in histological sections, cytospin preparations, or smears. Horseradish Peroxidase Avidin D is offered as a concentrate (5 mg/ml).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.