Learn More
Rockland Immunochemicals HORSE RED BLOOD CELLS (RBC) 10% Washed Pooled Cells - R409-0050
SDP
Supplier:  Rockland Immunochemicals R4090050
Encompass_Preferred
Horse whole blood is washed to remove the platelet rich plasma, buffy coat layer, and leukocytes (WBC). Red blood cells are supplied as a 10 percent suspension in phosphate buffered saline (PBS). Horse red blood cells are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Horse red blood cells are perishable and are collected and processed upon receipt of your order.
Catalog No. NC1525220