Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Horizontal Gel Box, 10 cm
SCI_ED

The Axygen™ HGB-10 is a compact 10 cm wide gel box capable of running up to 100 samples depending on configuration. It is ideal for quick sample-checks, particularly following restriction digestion during cloning.

Supplier:  Axygen™ HGB10/EMD

Catalog No. S28791

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View alternate product


Description

Description

The external gel casting provides efficiency as gel casting and gel running can occur simultaneously. The one piece injection molded tank is durable and leak free while the study electrode cassettes are user changeable. This compact unit is capable of separating bands over a distance of 9 cm. The system only requires 300 mL of buffer making it an economical choice. System includes the novel Axygen contrast enhancer that allows for easy visualization of wells for sample loading and is reversible with white background to visualize running of gels with colored MW markers.
  • Two size gel trays included 10x 10 cm and 10 x 12 cm
  • Axygen Contrast Enhancer
  • User replaceable electrodes
  • Low buffer volume
  • External gel casting
  • Recirculation ports
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE certified
300 mL
10 cm
Horizontal Gel Box
12.5 x 22 cm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit