Learn More
Honeywell Analytics™ GasAlertQuattro Multigas Detectors
$1486.00 - $2570.00
Specifications
Alarm Types | Audible, visual, vibrating |
---|---|
Battery Life | 14 hr. |
Battery Type | Alkaline |
Humidity Range | 10 to 100% |
Temperature Range (English) | -4°F to +122°F |
Catalog Number | Mfr. No. | Resolution | Components Measured | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Resolution | Components Measured | Color | Price | Quantity & Availability | |||
19-413-302
|
Honeywell Analytics™
QTXWHMABNA |
O2: 0.1%, LEL: 1%, H2S: 1ppm, CO: 1ppm | Carbon Monoxide, Hydrogen Sulfide, LEL, Oxygen | Black |
Each for $2,570.00
|
|
|||
19-413-306
|
Honeywell Analytics™
QTXW0MAYNA |
O2: 0.1%, LEL: 1%, CO: 1ppm | Carbon Monoxide, LEL, Oxygen | Yellow |
Each for $1,931.00
|
|
|||
19-413-305
|
Honeywell Analytics™
QTXWH0AYNA |
O2: 0.1%, LEL: 1%, H2S: 1ppm | Hydrogen Sulfide, LEL, Oxygen | Yellow |
Each for $1,931.00
|
|
|||
19-413-301
|
Honeywell Analytics™
QTXWHMAYNA |
H2S: 1ppm, CO: 1ppm, O2: 0.1 % v/v, LEL: 0.1 % v/v | Carbon Monoxide, Hydrogen Sulfide, LEL, Oxygen | Yellow |
Each for $2,560.00
|
|
|||
19-413-307
|
Honeywell Analytics™
QTX0HMAYNA |
O2: 0.1%, H2S: 1ppm, CO: 1ppm | Carbon Monoxide, Hydrogen Sulfide, Oxygen | Yellow |
Each for $1,931.00
|
|
|||
19-413-309
|
Honeywell Analytics™
QTXW00AYNA |
O2: 0.1%, LEL: 1% | LEL, Oxygen | Yellow |
Each for $1,777.00
|
|
|||
19-413-310
|
Honeywell Analytics™
QTX0H0AYNA |
O2: 0.1%, H2S: 1ppm | Hydrogen Sulfide, Oxygen | Yellow |
Each for $1,777.00
|
|
|||
19-413-312
|
Honeywell Analytics™
QTX00MAYNA |
O2: 0.1%, CO: 1ppm | Carbon Monoxide, Oxygen | Yellow |
Each for $1,777.00
|
|
|||
19-413-314
|
Honeywell Analytics™
QT00HMAYNA |
H2S: 1ppm, CO: 1ppm | Carbon Monoxide, Hydrogen Sulfide | Yellow |
Each for $1,777.00
|
|
|||
19-413-333
|
Honeywell Analytics™
QTXWH0ABNA |
O2: 0.1%, LEL: 1%, H2S: 1ppm | Hydrogen Sulfide, LEL, Oxygen | Black |
Each for $1,614.00
|
|
|||
19-413-334
|
Honeywell Analytics™
QTXW0MABNA |
O2: 0.1%, LEL: 1%, CO: 1ppm | Carbon Monoxide, LEL, Oxygen | Black |
Each for $1,931.00
|
|
|||
19-413-337
|
Honeywell Analytics™
QTXW00ABNA |
O2: 0.1%, LEL: 1% | LEL, Oxygen | Black |
Each for $1,777.00
|
|
|||
19-413-338
|
Honeywell Analytics™
QTX0H0ABNA |
O2: 0.1%, H2S: 1ppm | Hydrogen Sulfide, Oxygen | Black |
Each for $1,777.00
|
|
|||
19-413-335
|
Honeywell Analytics™
QTX0HMABNA |
O2: 0.1%, H2S: 1ppm, CO: 1ppm | Carbon Monoxide, Hydrogen Sulfide, Oxygen | Black |
Each for $1,931.00
|
|
|||
19-413-342
|
Honeywell Analytics™
QT00HMABNA |
H2S: 1ppm, CO: 1ppm | Carbon Monoxide, Hydrogen Sulfide | Black |
Each for $1,777.00
|
|
|||
19-413-340
|
Honeywell Analytics™
QTX00MABNA |
O2: 0.1%, CO: 1ppm | Carbon Monoxide, Oxygen | Black |
Each for $1,486.00
|
|
|||
Description
Specifications
Audible, visual, vibrating | |
Alkaline | |
-4°F to +122°F | |
11.92 oz. |
14 hr. | |
10 to 100% | |
-20°C to +50°C | |
338g |