Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell Analytics™ GasAlertMax XT II Multigas Detectors
Monitor up to four hazardous gases with a robust, motorized pump for intelligent, remote sampling. The GasAlertMax XT II Multigas Detector is compact, comfortable, field tough, and ideal for confined sampling.
Specifications
Alarm Types | Audible, visual, vibrating |
---|---|
Depth (English) | 2 in. |
Depth (Metric) | 5.08 cm |
Display Type | LCD |
Height (English) | 5.12 in. |
Catalog Number | Mfr. No. | Color | Resolution | Components Measured | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Resolution | Components Measured | Price | Quantity & Availability | |||
19-413-371
|
Honeywell Analytics™
XTXWHMYNA |
Yellow | O2: 0.1%, CO: 1ppm, H2S: 1ppm, Combustible gases: 0.1%, LEL: 1% | Carbon Monoxide, Hydrogen Sulfide, LEL, Oxygen |
Each for $1,863.00
|
N/A | |||
19-413-374
|
Honeywell Analytics™
XTXW0MYNA |
Yellow | O2: 0.1%, LEL: 1%, CO: 1ppm | Carbon Monoxide, LEL, Oxygen |
Each for $1,975.00
|
N/A | |||
19-413-372
|
Honeywell Analytics™
XTXWHMBNA |
Black | O2: 0.1%, LEL: 1%, H2S: 1ppm, CO: 1ppm | Carbon Monoxide, Hydrogen Sulfide, LEL, Oxygen |
Each for $1,817.00
|
N/A | |||
19-413-373
|
Honeywell Analytics™
XTXWH0YNA |
Yellow | O2: 0.1%, LEL: 1%, H2S: 1ppm | Hydrogen Sulfide, LEL, Oxygen |
Each for $1,975.00
|
N/A | |||
19-413-375
|
Honeywell Analytics™
XTX0HMYNA |
Yellow | O2: 0.1%, H2S: 1ppm, CO: 1ppm | Carbon Monoxide, Hydrogen Sulfide, Oxygen |
Each for $1,975.00
|
N/A | |||
19-413-377
|
Honeywell Analytics™
XTXW00YNA |
Yellow | O2: 0.1%, LEL: 1% | LEL, Oxygen |
Each for $1,957.00
|
N/A | |||
19-413-378
|
Honeywell Analytics™
XT00HMYNA |
Yellow | H2S: 1ppm, CO: 1ppm | Carbon Monoxide, Hydrogen Sulfide |
Each for $1,870.00
|
N/A | |||
19-413-379
|
Honeywell Analytics™
XTX0H0YNA |
Yellow | O2: 0.1%, H2S: 1ppm | Hydrogen Sulfide, Oxygen |
Each for $1,999.00
|
N/A | |||
19-413-380
|
Honeywell Analytics™
XTX00MYNA |
Yellow | O2: 0.1%, CO: 1ppm | Carbon Monoxide, Oxygen |
Each for $1,957.00
|
N/A | |||
19-413-388
|
Honeywell Analytics™
XTXW0MBNA |
Black | O2: 0.1%, LEL: 1%, CO: 1ppm | Carbon Monoxide, LEL, Oxygen |
Each for $1,980.00
|
N/A | |||
19-413-387
|
Honeywell Analytics™
XTXWH0BNA |
Black | O2: 0.1%, LEL: 1%, H2S: 1ppm | Hydrogen Sulfide, LEL, Oxygen |
Each for $1,980.00
|
N/A | |||
19-413-389
|
Honeywell Analytics™
XTX0HMBNA |
Black | O2: 0.1%, H2S: 1ppm, CO: 1ppm | Carbon Monoxide, Hydrogen Sulfide, Oxygen |
Each for $1,980.00
|
N/A | |||
19-413-391
|
Honeywell Analytics™
XTXW00BNA |
Black | O2: 0.1%, LEL: 1% | LEL, Oxygen |
Each for $1,961.00
|
N/A | |||
19-413-392
|
Honeywell Analytics™
XT00HMBNA |
Black | H2S: 1ppm, CO: 1ppm | Carbon Monoxide, Hydrogen Sulfide |
Each for $1,872.00
|
N/A | |||
19-413-393
|
Honeywell Analytics™
XTX0H0BNA |
Black | O2: 0.1%, H2S: 1ppm | Hydrogen Sulfide, Oxygen |
Each for $2,010.00
|
N/A | |||
19-413-394
|
Honeywell Analytics™
XTX00MBNA |
Black | O2: 0.1%, CO: 1ppm | Carbon Monoxide, Oxygen |
Each for $1,961.00
|
N/A | |||
Description
- Integrated sampling pump with reliable diaphragm technology
- Full function self-test of sensor, battery status, circuit integrity and audible/visual alarms on start up
- Equipped with standard datalogging and event logging
- Continuous LCD shows real-time gas concentrations
- Block detection with semi-conductor pressure sensor
- User adjustable alarm setpoints
- Multi-language support
- Water-resistant
- Size: 13.1 x 7.0 x 5.2cm (5.1 x 2.8 x 2.0 in.)
- Weight: 316g (11.1 oz.)
- Temperature Range: -20° to +50°C (-4° to +122°F)
- Humidity: 10 to 100% RH (non-condensing)
H2S Sensors
- Measuring Range: 0 to 200ppm
- Resolution: 1.0ppm
- Measuring Range: 0 to 1000ppm
- Resolution: 1.0ppm
- Measuring Range: 0 to 30% by vol
- Resolution: 0.1%
- Measuring Range: 0 to 100% LEL; 0 to 5.0% v/v
- Resolution: 1.0% LEL; 0.1% v/v
Warranty and Services
Full 2 year warranty including all sensors
Specifications
Audible, visual, vibrating | |
5.08 cm | |
5.12 in. | |
10 to 100% | |
-20°C to +50°C | |
316g | |
7.2 cm |
2 in. | |
LCD | |
13 cm | |
-4°F to +122°F | |
11.1 oz. | |
2.8 in. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title