Learn More
Hoefer™ Mylar™ Casting Tape
SDP
Ideal for sealing glass plate sandwiches when casting polyacrylamide gels (especially manual sequencing gels), or for sealing the ends of running trays when casting agarose gels
Supplier:  Hoefer™ SE1510
Catalog No. 03-500-468
  • $30.98 / Each of 1
    Save 69%
    Reg : $99.00
    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
    Learn More
Add to cart
This item is not returnable. View return policy
Description
Description
  • For use with SE900 Large Format Vertical Gel Protein Electrophoresis Unit
Specifications
Specifications
Ideal for sealing glass plate sandwiches when casting polyacrylamide gels or for sealing the ends of running trays when casting agarose gels
65.8 mm
Mylar Casting Tape
2.5 cm