Learn More
PIP™ HL Bouton Softside Goggle
Click to view available options
Lens Tint:
Clear
Lens Coating:
Scratch-resistant
Description
- Solid, non-ventilated frame
- Soft vinyl body molds snugly to the face without pressure or irritation
- Fits comfortably over most prescription glasses
- Extended face flanges close the gaps over the nose and cheekbone areas, providing excellent protection
- Clear blue transparent frame to prevent yellowing
- Clear, crisp and distortion free visibility while meeting the requirement standards that help provide protection from projectiles and impact in general use applications
- Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching
- Anti-fog coating eliminates the condensation of moisture on lenses that causes fogging so your lenses and vision stay clear
Specifications
Lens Coating | Scratch-resistant |
Lens Tint | Clear |
Product Type | Goggles |
For Use With (Application) | Provides protection from chips and flying particles |
Ventilation Details | Non-vent |
UV Protection | 0.999 |