Learn More
Sigma Aldrich Fine Chemicals Biosciences Histopaque-1077, 6 x 100 mL
Supplier: Sigma Aldrich Fine Chemicals Biosciences 107716X100ML
Histopaque-1077 is a sterile, endotoxin tested solution of polysucrose and sodium diatrizoate, adjusted to a density of 1.077g/mL. This ready-to-use medium facilitates rapid recovery of viable lymphocytes and other mononuclear cells from small volumes of whole blood.