Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ HiPur™ 10% Neutral Buffered Formalin
Choose the option that best fits your workflow, HiPur™ 10% neutral buffered formalin is available in one and five gallon sizes.
$107.00
Specifications
Color | Colorless |
---|---|
Content And Storage | Room temperature |
Concentration | 0.1 |
For Use With (Application) | Histological fixative to fix tissues |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
22-110-873
|
Epredia™
94001 |
4 x 1 gal. |
Case of 4 for $132.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-110-875
|
Epredia™
94005 |
5 gal. |
Each for $107.00
|
|
|||||
Specifications
Colorless | |
Room temperature | |
0.1 | |
Histological fixative to fix tissues |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title