Learn More
Fischer Technical Company High Vacuum Exhaust Filter
High Vacuum Exhaust Filter. 0.5 in. Brass elbow.
Supplier: Fischer Technical Company LAVEF
Alert:
Prop 65
Description
- High Vacuum exhaust filter to be used with the LAV series High Vacuum pumps.
- The 0.5 in. Brass elbow fits on the end our LAV-Series High Vacuum Pump handles which are also the exhaust for the pump.
Certifications
SciEd
Specifications
High Vacuum | |
LAV Series High Vacuum Pumps |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov