Learn More
Fisherbrand™ High Precision #10 Style Scalpel Blade
For use in various applications. Fisherbrand High Precision #10 Style Scalpel Blade is made of stainless steel and packaged in individual foil packs.
Description
- Features curved cutting edge
- Works with #3 scalpel handle
- Replacement blade for PL-DIS-2
Specifications
Stainless Steel | |
# 10 | |
Scalpel (Blade Only) |