Learn More
Amerex™ High-Performance Fire Extinguisher with ABC Fast Flow
Amerex™ High-Performance Fire Extinguishers are available with a proprietary, zinc-enriched coating that resists corrosion for up to 12 years in severely salt-contaminated environments.
Supplier:  Amerex™ 26905
 View more versions of this product
Catalog No. 19-860-568
Add to cart
Description
Description
  • Features zinc enriched primer coating
  • Resists extreme environmental conditions
  • Highest achievable UL ratings
  • Proprietary corrosion-resistant paint process
  • Simple operation and maintenance
  • Improved environmental impact compared to galvanized units
  • ABC Fast Flow
Specifications
Specifications
Fire Extinguisher
Stainless Steel, Brass, Steel
13.61 kg
21 sec.
792
4A:40B:C
26.3 kg
Red
30 lb.
8 x 11 x 26.5 in. (20.32 x 27.94 x 67.31 cm)
Wall Hanger, Protective Foot Ring
40 to 50 ft.
58 lb.