Learn More
Pyramex™ Hi-Vis Mesh Safety Vests
SDP
Catalog Number p-7182992
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Pyramex™ Hi-Vis Mesh Safety Vests with ANSI/ISEA 107-2015 safety compliance
  • Front: Hook & Loop closure
  • One 2 in. silver reflective stripe applied horizontally 8.5 in. from the bottom
  • Two 2 in. silver reflective stripes run vertically from top of horizontal stripes, over shoulders to top of back side horizontal stripe
Closure Type Hook and Loop
Certifications/Compliance ANSI/ISEA 107-2015, Type R - Class 2
Color Hi-vis Lime
Size Small
Material Polyester Mesh
Size (English) Chest 34 to 36 in.
Tape Type China Brand Flat Silver