Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HF Scientific™ ProCal™ Turbidity Standard Reagents Kit for Hach™ 2100P Portable Turbidimeter
Standards not harmful to the environment or to worker safety
Specifications
For Use With (Equipment) | Hach 2100P Portable Turbidimeter |
---|---|
Certifications/Compliance | EPA |
Catalog Number | Mfr. No. | Type | Turbidity | Volume (English) | Volume (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Turbidity | Volume (English) | Volume (Metric) | Price | Quantity & Availability | ||
15-393-625
|
HF Scientific™
52390 |
Primary Turbidity Standard Kit | 0, 20, 100, 800 NTU | 8.45 oz. | 250 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
15-393-628
|
HF Scientific™
H52420 |
Primary Turbidity Standard | 0 NTU | 16.9 oz. | 500 mL | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||
15-393-629
|
HF Scientific™
H52430 |
Primary Turbidity Standard | 20 NTU | 8.45 oz. | 250 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
Description
- Pre-diluted
- Do not include formazin
- Preservative used enhances stability
- Biodegradable, safe and non-toxic
- Ready and easy to use
- Factory certified by HF scientific
- Custom NTU produced
- Available in different size bottles
Specifications
Hach 2100P Portable Turbidimeter | |
EPA |
Safety and Handling
ShelfLife : 2 Years
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title