Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herbicides, MilliporeSigma™ Supelco™

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Supplier:  MilliporeSigma Supelco QC11242ML

Catalog No. 11-101-7362


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

Featured Industries: Agriculture and Environmental
Specifications

Specifications

1634-04-4
2 mL
Certified Reference Material
Certified Reference Material
-33°C - closed cup - Solvent
Safety and Handling

Safety and Handling

Recommended Storage : 2°C to 8°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit