Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heptane, HPLC Grade, Spectrum™ Chemical

Click to view available options
Quantity:
1 L
4 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Description
Spectrum™ Chemical Heptane, HPLC Grade is a straight-chain alkane applied in laboratories as a non-polar mobile phase carrier solvent in high pressure liquid chromatography. All HPLC solvents offered by Spectrum are glass distilled, submicron filtered, and undergo rigorous specification testing to insure accurate, repeatable results.
Specifications
CAS Max % | 100% |
UN Number | UN1206 |
Grade | High Purity Solvent |
Quantity | 6 x 1 L |
Packaging | Amber Glass Bottle |
Residue after Evaporation | 2 ppm |
Water | 0.02% |