Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heptane, HPLC Grade, Spectrum™ Chemical
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Description
Spectrum™ Chemical Heptane, HPLC Grade is a straight-chain alkane applied in laboratories as a non-polar mobile phase carrier solvent in high pressure liquid chromatography. All HPLC solvents offered by Spectrum are glass distilled, submicron filtered, and undergo rigorous specification testing to insure accurate, repeatable results.
Specifications
CAS Max % | 100% |
UN Number | UN1206 |
Grade | High Purity |
Quantity | 1 L |
Packaging | Amber Glass Bottle |
Residue after Evaporation | 2 ppm |
Water | 0.02% |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction