Learn More

Heparin, Fisher BioReagents
Description
Specifications
Color | White |
pH | 5.0 to 7.5 |
Loss on Drying | ≤5.0% |
Packaging | Poly Vial |
Merck Index | 15, 4688 |
Quantity | 50 mg |
Formula Weight | ≈5000 |
Vapor Pressure | Negligible |
Physical Form | Powder or Solid |
Chemical Name or Material | Heparin, From Porcine Intestinal Mucosa |
Safety and Handling

CAUTION!
Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. The toxicological properties have not been fully investigated. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:0
Instability:0
Recommended Storage : RT
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.