Learn More
Thermo Scientific™ HEPA Filter Kit
For Use With: Steri-Cult 3307/3308, 3310/3311 Incubators
Supplier:  Thermo Scientific™ 1900160
Catalog No. 13-998-104
Add to Cart
Description
Description
Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
HEPA Filters
  • 99.97% filtering at 0.3μm
  • Compatible with Thermo Scientific™ Forma™ Steri-Cult™ 3307 and 3310 CO2 Incubators
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Decontamination Kits
  • Consists of filters for difficult-to-clean chambers and gaskets (except door) and tubing
  • Reduces downtime and labor spent on decontamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HEPA Filter Replacement Kit
Steri-Cult™ 3307 and 3310 CO2 Incubators
LED
HEPA Filter
Product Suggestions
Product Suggestions