Learn More
EUROTROL HEMOLIN 5 VIALS/BOX
Supplier: EUROTROL 023.000.005
A 5 level linearity panel to validate the entire reportable range for hemoglobin on HemoCue Hb201 systems. Formulated with real hemoglobin. Features 30 day open vial stability at room temperature. Validated by HemoCue.