Learn More
Newcomer Supply Helicobacter sp. Control Slides, 10/set
Supplier: Newcomer Supply 4276A
Newcomer Supply Helicobacter sp. Control Slides are for the positive histochemical staining of Helicobacter sp., a spiral shaped bacterium associated with stomach inflammation and implicated in the development of gastric malignancy, peptic ulcers, and chronic gastritis. Helicobacter sp. Control Slides contain sections of positive staining gastrointestinal tract.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.