Learn More
Invivogen HEK-Blue™ hTLR8, 3-7 x 10e6 cells
Supplier: Invivogen HKBHTLR8
HEK-Blue™ hTLR8 cells were derived from the human embryonic kidney HEK293 cell line to monitor the activation of human TLR8. The endosomal location of TLR8 allows it to scan for the presence of microbial RNA in the phagocytic cargo.