Learn More
OHAUS™ Heavy Duty Shaker
SCI_ED
OHAUS Heavy Duty Shakers are designed to handle a range of applications. Digital model with microprocessor control to provide variable speed and consistent shaking, while ramping to the set speed. Built-in tray and non-slip rubber mat included.
Manufacturer:  OHAUS™ 30391807/EMD
Catalog No. S72071
Add to cart
Description
Description
  • Exceptional speed control, accuracy and durability
  • LED displays for speed and time
  • Calibration mode for speed
  • Overload protection and speed ramping feature
Specifications
Specifications
19 mm
110 V
35 lb.
13 in.
33 cm
27.9 cm
SHHD1619DG
15 to 500 rpm
120 V
14.9 cm
41.3 cm
35.5 cm
Digital
Non-skid rubber mat and Power cord