Learn More
Chemglass Life Sciences Glas-col Heating Tape, 1in Wide x 5ft Long, 100 Watts, 115 Volts, 27.25 Ohms/ft
SDP
Supplier:  Chemglass Life Sciences CG1405018
Encompass_Preferred
Glas-col Heating Tape, 1in Wide x 5ft Long, 100 Watts, 115 Volts, 27.25 Ohms/ft. Tape heats odd lengths of glass or metal pipe. Ribbon-type heating elements running lengthwise in the tape are encased in durable glass braid and are impregnated and covered with tough silicone rubber. The silicone rubber insulates stranded terminal leads at one end & waterproofs the fittings at both ends. It also permits use in high-moisture areas & helps to make the tapes resistant to most laboratory environment conditions. All tapes are supplied with a 2 blade plug and have a temperature limit of 480ºF. NOTE: Power ratings listed are for tapes not covered by insulation when installed. If covered, temperature should be monitored & controlled to avoid exceeding the 480ºF limit. Short narrow tapes that are rated at less than 115 volt must be used with a variable voltage transformer.Due to the voltage of the heating tape, a variable voltage transformer is required. Please see CG15005, CG15006 or CG15010.
Catalog No. CG1405018