Learn More
Cole-Parmer A.I.E AIE-300C Heat Sealer with Built-In Cutter, 12" , 115 V
Supplier: Cole-Parmer 0301830
This heat sealer produces up to 12" long seals, and the cutter slices away excess material 1/4" (6.4 mm) from the 5/64" (2 mm) wide seal. Maximum material thickness for proper sealing is 6 mil.