Learn More
Chemglass Life Sciences Heat Gun, Heavy-Duty, Multiple Use, 649°C Temp, 120V, 60Hz, 14.5A, 1740W
SDP
Supplier:  Chemglass Life Sciences CG9001D03
Encompass_Preferred
Professional, heavy-duty, multiple use heat gun has a sturdy die cast housing and adjustable non-slip stand. Used to bend plastics, cure adhesives, dry parts, heat parts, shrink tubing and packaging, strip paint and more. Heat gun accommodates many industrial and commercial heating applications.
Catalog No. NC1808422