Learn More
Victory Innovations™ Handheld Electrostatic Sprayer
Handheld Electrostatic Cordless Sprayer is ideal for disinfecting, sanitizing, odor removal, decontamination, and more. Save time and money and get the best chemical coverage for the job.
Supplier: Victory Innovations™ VP200ESK
Description
Victory Innovations™ Handheld Electrostatic Cordless Sprayer is ideal for disinfecting, sanitizing, odor removal, decontamination, and more. Save time and money and get the best chemical coverage for the job.
- 3-in-1 nozzle allows flexibility in spraying
- 40-micron, 80-micron, and 110-micron settings to match chemical dwell times
- Covers 2800 square feet on 40-micron nozzle setting
- Rechargeable lithium-ion battery
- 4 hours of continuous spray time
Specifications
Lithium-ion Battery | |
Disinfecting, odor removal and decontamination | |
3-in-1 Nozzle | |
Handheld Sprayer |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.