Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laxco™ Handheld Analog Clinical Refractometer

When you need reliable, accurate readings of your Urine samples, the Laxco™ Analog Industrial Clinical Refractometers rugged design is built to last. With the ability to read Serum Protein, Specific Gravity and Refractive Index all on a single scale, it makes it the ideal choice.

Supplier:  Laxco™ IRC200

 View more versions of this product

Catalog No. IRC200

Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. View substitute products »


Includes: Calibration Screw Driver, Hard Sided case, Applicator, Calibration fluid, Manual

Description

Description

Refractometers have been used for measuring the concentration of solutions for more than a century. Their primary purpose is to measure the degree at which light bends as it passes through a medium, and is used in a wide range of applications and industries as a quality control and diagnostic tool.

Laxco Analog Refractometers are designed for simple operation and built with all a metal construction for durability. Its industrial design is the most commonly used design among refractometer manufactures as it is a cost-effective model to build.

Each unit is inspected in the United States by a factory qualified technician under a rigid quality control process to ensure its accuracy, and performance.

Key Features:

  • Quickly calibrate the instrument with a turn of a screw, and distilled water
  • Comfortable eyecup blocks out ambient light and allows for crystal clear viewing of readings
  • Automatic temperature compensation prevents the need for constant re-calibration of the instrument. As the ambient temperature changes, the scale will automatically compensate for the difference in the degree of refraction. This ensures an accurate reading every time
  • All-metal construction creates durability. NOTE: Model 12-561-340 comes without ATC
  • Footrest allows the instrument to be set on a flat surface without the worry of it rolling off working surfaces
  • Rubber hand grip is comfortable, easy to grasp. Its non-slip pattern grip reduces that chance of accidental dropping the instrument
  • Double hinged daylight plates create two points of movement to allow for greater flexibility in movement; making it easy to add samples and clean
  • Portable, easy to use design.

Clinical Models:

Quickly and accurately get an indication of basic health parameters such as dehydration and blood plasma protein levels. Laxco Analog Clinical Refractometers uses three-scales to measure Total Serum Protein, Urine Specific Gravity, and Refractive Index. They are ideal for use in quality control, research, and clinical laboratories.

  • Urine Specific Gravity: 1.000ppt to 1.050ppt with granularity to measure down to 0.001ppt at a 0.002ppt degree of accuracy
  • Serum Protein: 1g to 12g with granularity to measure down to 0.2g/100mL at a 0.2g/100mL degree of accuracy
  • Refractive Index: 1.334nD to 1.360nD with granularity to measure down to .0005nD at a 0.0005nD degree of accuracy.

Warranty and Services

5 Years

Specifications

Specifications

Clinical Refractometer
1.331 to 1.36
Distilled Water
0.0005
Linear
Clinical
IP54
0.881 lb.
172 x 39 x 39 mm
39 mm
204 mm
39 mm
5 years optical and mechanical, 1 year electrical from date of purchase
1.000 to 1.050 Urine Specific Gravity, 1 to 12 g/dl Serum Protein
1 mL
0.002; 0.2 g/dl; 0.0005
0.002; 0.2 g/dl; 0.0005
Ambient
Calibration Screw Driver, Hard Sided Case, Applicator, Calibration Fluid, Manual
1
0.4 kg
1.535 in.
6.772 in.
1.535 in.
No
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Warranty: Does not apply to daylight plate assembly