Learn More
LabScientific Halo-Carbon Oil # 700, injection work, 8 oz. Safe, non-flammable, chemically inert.
Supplier: LabScientific FLY7000
Halo-Carbon Oil # 700, injection work, 8 oz. This oil is safe, nonflammable and chemically inert – safe for oxygen, chlorine and most reactive gas services. It has high viscosity, good lubricity, high thermal stability, and low compressibility.