Learn More
Honeywell Miller™ H500 Series Safety Harness, Model CC1
The modern ergonomic design provides high visibility and smart features, in addition to safe and comfortable functionality. Available in various models, each in three different sizes.
Supplier: Honeywell Miller™ H5CC311022
Description
- Lightweight, ergonomic, breathable harness
- Comfortable around the legs, waist, back and shoulders
- Smart rebar hook/lanyard keepers and SRL web link
- Increased freedom of movement
- Quick and easy-to-fit and adjust
- Improved visible stitch pattern
- Clearly identifiable fall-impact indicators
Specifications
Universal | |
H500 CC1 UNIV STL TB/MB SD | |
Fall Protection Harness |
Tongue/Mating | |
Side | |
Steel |