Learn More
H-B Instrument™ Minimum/Maximum Glass Bimetal Thermometer
SCI_ED
Reset thermometer with easy-to-use push button.
Supplier:  H-B Instrument™ B607610000
Catalog No. S02697
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
  • Measures both minimum and maximum temperatures in Fahrenheit and Celsius scales
  • Readings are accurate to ±1 scale divisions
  • Bulbs are enclosed in a sturdy plastic case suitable for outdoor or indoor use
Specifications
Specifications
High moisture or wet environments including green houses, sample preparation areas and water treatment facilities
120°F
-30°F
-30°F to +120°F
1 at Ambient, 2 at -30°C tolerance between limbs 1.5
Glass
50°C
-35°C
-35°C to +50°C
1°C/1°F
Product Suggestions
Product Suggestions