Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-B Instrument™ Durac™ Plus™ Blue Spirit Certified Glass Thermometers
SCI_ED

Ideal for use in applications where a very precise accurate temperature measurement is required.

Supplier:  H-B Instrument™ B602010100

Catalog No. S02666

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

  • Feature precise and accurate temperature measurement
  • Nonmercury blue spirit liquid
  • Triangular anti-roll fitting, permanently fused markings, blue spirit liquid, white background glass
  • Individually serial numbered
  • Length: 16.5 in. , with 3 in. immersion
Specifications

Specifications

0, 37 and 56°C
16.5 in.
Glass
0.1°C
ISO/IEC 17025:2005, A2LA, NIST
41.9 cm
1°C to 61°C
3 in.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit