Learn More
Rockland Immunochemicals GUINEA PIG COMPLEMENT (lyophilized) with DILUENT - C200-0005
Supplier: Rockland Immunochemicals C2000005
Special processing techniques are used to yield products with high complement activity and low background cytotoxicity. Guinea Pig Complement is suitable for CFT and SRH.