Learn More
Thermo Scientific GUARD COLUMN
Supplier: Thermo Scientific 056899
IonPac AG14A-5 micrometer Guard Column, Dimensions: 3 x 30 mm,Maximum Operating Pressure: 4000 psi, Mobile Phase Compatibility: pH 0 -14;0 -100%HPLC solvents,Pore Size:100 angstrom, Crosslinking (%DVB): 55%, Hydrophobicity:Moderate