Learn More
GSC Go Science Crazy Support Stand with Rings
Great for holding all sorts of lab equipment in a small space. The GSC Support Stand with Rings is made from cast iron, with an enamel finish that is acid resistant. Includes a zinc plated steel rod that screws into the base support. Supplied with support rings which are also cast-metal with a zinc finish.
Supplier: GSC Go Science Crazy 4SSC15R3
Alert:
Not a toy. Product intended for ages 13 and up.
Includes: Includes cast metal support rings with zinc finish
Description
- Stand made from cast iron
- Acid resistant
- Plated steel support rod
- Rod threaded to fit base
- Includes support rings
Specifications
3-Ring |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov