Learn More
GSC Go Science Crazy Sintered Glass Funnels
Quality borosilicate glass funnel with porous filter plate.
Supplier: GSC Go Science Crazy FNLSINT1253
Description
- Removable porous filter plate for cleaning and reuse
- Allows fine filtration without use of filter paper
Specifications
125 mL | |
Borosilicate Glass | |
3, Fine |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov