Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Grips™ Premium Goatskin Leather Drivers Gloves

Catalog Number p-145684
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Goatskin leather is soft, pliable, and longer lasting than cow and pig leather drivers. Grips™ Premium Goatskin Leather Drivers Gloves are ideal for use in warehousing, construction, fork lift driving, and oil and gas industries.

  • Top quality goatskin with natural added lanolin for softness and extended wear
  • Goatskin has a higher natural tensile strength despite abrasion
  • Gunn cut and keystone thumb for added dexterity and flexibility
  • Bound hem
Color Beige
Grade Premium
Hand Specific
Material Goatskin Leather
Product Type General Purpose Gloves
Size Small
Style Gunn
Glove Cuff Style Open
Hem Color Red
Lining Unlined
Series Y0769
Thumb Style Keystone
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit