Learn More
Sarstedt Inc GRIPPER RACK 18 PLACE SCMTUBES
Supplier: Sarstedt Inc 93.894
Gripper Rack; 18 Place for Screw Cap Microtubes; Polycarbonate; 10.5mm; suitable for micro tubes with screw cap; notched aperature enables single-handed operation