Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Grasshoppers

Brachystola sp.
$16.00 - $66.50
Catalog Number | Mfr. No. | Quantity | Injection Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Injection Type | Price | Quantity & Availability | ||||
S8538010
![]() |
Ranaco Corporation
126.1881.10 |
10 Pk. | Single |
Pack of 10 for $16.00
|
|
||||
S4403S
![]() |
Ranaco Corporation
126.1873.10 |
10 Pk. | Plain |
Pack of 10 for $7.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S1875S50
![]() |
Ranaco Corporation
126.1873.50 |
50 Pk. | Plain |
Pack of 50 for $33.00
|
|
||||
S1875S100
![]() |
Ranaco Corporation
126.1873.100 |
100 Pk. | Plain |
Pack of 100 for $66.50
|
|
||||
Description
- Lubber grasshoppers we use are all large, “Grade A” specimens
- Each internally injected with a special formulation that ensures flexibility and preservation
- Single-injected grasshoppers have their tracheal systems filled with colored latex
- Size: 3 in. (7.6cm)
Specifications
Grasshopper | |
3 in. | |
7.6 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title