Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Graham-Field™ Kelly Hemostatic Forceps

For medical and surgical procedures
Specifications
Product Type | Kelly Hemostatic Forcep |
---|---|
Material | Stainless Steel |
For Use With (Application) | For medical, surgical procedures |
Description
- Stainless steel
- 5.5 in. (14cm)
- Available straight or curved
Specifications
Kelly Hemostatic Forcep | |
Stainless Steel | |
For medical, surgical procedures |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title