Learn More
Promega GoTaq MDx DNA Polymerase, Glycerol-Free, 500u
GoTaq MDx DNA Polymerase is a full-length form of Taq DNA polymerase that exhibits 5'→3' exonuclease activity.
Supplier: Promega D4101
This item is not returnable.
View return policy
Description
- Full−Length Form of Taq DNA Polymerase Manufactured Under cGMP
- Glycerol−free formulation available
- Custom formats and formulations
- Supplied at a concentration ≥5u/microl
Specifications
-30°C to -10°C | |
500 U |