Learn More
Ted Pella Inc Gold Colloids (Sols) | 5nm | 20mL
Supplier: Ted Pella Inc 1570220
Nanometer-sized gold particles of uniform shape and size are invaluable tools in nanotechnology (assembled arrays), light scattering (elastic - plasmon resonance; inelastic - Raman effect) and for single molecule detection. Gold nanoparticles can be attached to proteins, alkanethiols and DNA by various methods. Silver nanoparticles are evolving similar functional characteristics to their gold counterparts.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.