Learn More
Tinius Olsen Testing Machine Go/No-Go Gauge, standard ASTM D 1238/ISO1133 orifice
Supplier: Tinius Olsen Testing Machine 02001034
The Go/No-Go gauge is used to check the inner diameter of the standard ASTM D 1238 orifice (die) for compliance to the test method requirements.