Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyphosate Stock Standard, SPEX CertiPrep™

Click to view available options
Quantity:
1mL
Packaging:
2mL amber prescored glass ampule
1071-83-6, 7732-18-5
C3H8NO5P
169.07
MFCD00055350
XDDAORKBJWWYJS-UHFFFAOYSA-N
3496
CHEBI:27744
2-[(phosphonomethyl)amino]acetic acid
OC(=O)CNCP(O)(O)=O
1071-83-6, 7732-18-5
C3H8NO5P
169.07
MFCD00055350
XDDAORKBJWWYJS-UHFFFAOYSA-N
3496
CHEBI:27744
2-[(phosphonomethyl)amino]acetic acid
OC(=O)CNCP(O)(O)=O

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Glyphosate1071-83-62000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

CAS Min % 0.2, 99.8
CAS Max % 0.2, 99.8
Packaging 2mL amber prescored glass ampule
Methods For US EPA Methods 535, 547
Solvent or Matrix H2O
Quantity 1mL
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Chemical Name or Material Glyphosate Stock Standard, USEPA Method 547

DOTInformation : Nonhazardous

Recommended Storage : Ambient

Product intended for laboratory use only