Learn More
Miles Scientific Glass Reagent Sprayer
Supplier: Miles Scientific A4100
A glass sprayer with a specially designed head to eliminate formation of droplets and spattering. This feature assures delivery of a fine misty spray and ends problems from an uneven discharge. Can be used with an air-line or the rubber bulb supplied. The volume of the reagent flask is 100 ml.