Learn More
Leica Biosystems GLASS,PEN-MEMBRANE SLIDES 50CS
Supplier: Leica Biosystems 11505189
These slides are equipped with wells that are specifically designed for the laser microdissection of living cells. Each slide has 18 recesses with a PEN membrane base. Ibidi slides can be combined with the entire range of collectors suitable for the LCC mode. Another contamination-free selection variant are so-called slide stacks (Figures 11a, 11b), consisting of 2 Ibidi slides arranged on top of each other in sandwich technique. Cells dissected from such a stack fall by the force of gravity straight into the next lower Ibidi slide, which is fixed in a special stack holder (Figures 11a, 11b). Subsequent recultivation is possible as well as the downstream analysis of the extracted cells. A matching cap offers a further contamination protection option.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.